Chem Snap Cap, Clear with Molded Septum, Size: 8mm
Catalog #: 75820-760
Supplier: Chemglass
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Potassium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, , VWR® BDH®
Supplier: VWR International
$338.38