Aluminum Seal Red 11mm
Catalog #: 75820-750
Supplier: Chemglass
Customers Who Bought This Also Bought
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
$179.52 to $927.60
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Supplier: VWR International
$81.84 to $877.92
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$135.75 to $468.77
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$885.46 to $3,541.82
Iron, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
$415.20