Aluminum Seal Silver 20mm
Catalog #: 75820-720
Supplier: Chemglass
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Sodium hydroxide 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$72.65 to $846.50
Fluoride Standard, 1 ml = 0.1 mg F-, 100 ppm F-, Ricca Chemical Company
Supplier: Ricca Chemical
$162.76