Aluminum Seal Gold 11mm
Catalog #: 75820-718
Supplier: Chemglass
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Reagent alcohol absolute 94.0-96.0%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$356.35 to $1,425.43
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$741.97 to $47,885.70
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
$90.42 to $94.61