Insert, 0.25mL, Glass, BM with Glass Flange. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. Of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
Catalog #: 75820-696
Supplier: Chemglass
Customers Who Bought This Also Bought