Storage Rack for Vials, Polyethylene, Chemglass
Catalog #: 75820-618
Supplier: Chemglass
Specifications
- Material:Polyethylene
- No. of Holes:50
- Cat. no.:75820-618
- For Tubes:12×32 mm
- Supplier No.:CV-6005-0012
Specifications
About this item
Ordering information: Vials and closures must be purchased separately.
Customers Who Bought This Also Bought