De-Capper, Pliers, Hand Operated, Size: 11mm
Catalog #: 75820-610
Supplier: Chemglass
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Supplier: Medicom
$49.54 to $495.19
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
VWR® PBS-T (Phosphate Buffered Saline - Tween-20) Buffer Powder Packs
Supplier: VWR International
$212.19
Cotton Tipped Applicator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$23.40 to $26.50