Preassembled Closure and Septa GPI 8-425 Black
Catalog #: 75820-568
Supplier: Chemglass
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
214,10 $ to 1 039,32 $
Acetate Buffer Solution, pH 4.0, VWR Chemicals BDH®
Supplier: VWR International
270,18 $ to 1 066,09 $
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, sterile
Supplier: Teknova
386,34 $
Potassium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
830,89 $ to 3 323,50 $