Poly Crimp* Seal Yellow 11mm
Catalog #: 75820-558
Supplier: Chemglass
Customers Who Bought This Also Bought
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52
VWR® ARISTAR® Multi-Element ICP and ICP-MS Certified Reference Standards, Enhanced Packaging
Supplier: VWR International
$154.78
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
Supplier: Ricca Chemical
$150.16 to $1,502.77
Richter Anti-Vibration Pad, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$99.05 to $109.04
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$39.55