Cap Only 18mm Headspace Magnetic
Catalog #: 75820-480
Supplier: Chemglass
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
$971.75 to $3,463.97
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44
Potassium hydroxide 10% (w/v) in aqueous solution, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$64.23 to $385.27
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) 97%
Supplier: Thermo Scientific Chemicals
$155.31
Sodium dihydrogen phosphate monohydrate 99.0-102.0% (by alkalimetry), EMSURE® ACS for analysis, Supelco®
Supplier: MilliporeSigma
$335.36 to $1,972.67