ContactUsLinkComponent
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Catalog #: 75820-412
Supplier:  Chemglass
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Vial, 2.0mL, Amber with White Grad Spot, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Catalog #: 75820-412
Supplier:  Chemglass
Customers Who Bought This Also Bought