ContactUsLinkComponent
Vial, 1.5mL, Clear, Large Opening, 12x32mm, 9mm Thread, High Recovery. With a 40% larger opening, this vial is specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Catalog #: 75820-378
Supplier:  Chemglass
Vial, 1.5mL, Clear, Large Opening, 12x32mm, 9mm Thread, High Recovery. With a 40% larger opening, this vial is specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Vial, 1.5mL, Clear, Large Opening, 12x32mm, 9mm Thread, High Recovery. With a 40% larger opening, this vial is specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial.
Catalog #: 75820-378
Supplier:  Chemglass
Customers Who Bought This Also Bought