Western Blotting Kit, Azure Biosystems
Catalog #: 75818-196
Supplier: Azure Biosystems
Product available on GSA Advantage®
Specifications
- Description:AzureSpectra RG Western Kit, with Fluorescent Block
- Size:10 Blots
- Cat. no.:75818-196
Specifications
About this item
Azure's fluorescent western kits provide reagents and materials for two-color fluorescent Western blots.
- Pre-cut membranes
- Wash buffer
- Visualize two proteins by fluorescence
Western Blotting Kit for visualizing two proteins by fluorescence. Enough for 10 Western blots. Each kit includes pre-cut membranes, wash buffer, fluorescent blot blocking buffer, quenching sheets, and secondary antibodies.
Customers Who Bought This Also Bought