Order Entry
ContactUsLinkComponent
Standard Methods Agar (Plate Count Agar), 500ml
Standard Methods Agar (Plate Count Agar), Hardy Diagnostics
Catalog #: 75814-008
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Standard Methods Agar (Plate Count Agar), Hardy Diagnostics
Catalog #: 75814-008
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Standard Methods Agar
  • Size:
    Pack of 10
  • Volume:
    500 ml
  • Supplier No.:
    U297

Specifications

About this item

Standard Methods Agar is used for the cultivation and enumeration of microorganisms in dairy products, food, and water.

  • Cultivate and enumerate microorganisms

For the cultivation and enumeration of microorganisms in dairy products, food, and water.

Customers Who Bought This Also Bought