Order Entry
ContactUsLinkComponent
Blank disks, 0.25 inch diameter paper disk for AST
Blank Paper Disk for AST, Hardy Diagnostics
Catalog #: 75813-972
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
Blank Paper Disk for AST, Hardy Diagnostics
Catalog #: 75813-972
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Blank Disk
  • Size:
    0.25"
  • Cat. no.:
    75813-972
  • Supplier No.:
    Z7121

Specifications

About this item

A blank paper disk that can be used for inhibition studies.

  • Prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing
  • Used for inhibition studies

A blank paper disk that can be used for inhibition studies. Used to prepare your own disks for Kirby Bauer (antimicrobial disk diffusion (AST)) testing on Mueller Hinton Agar.

Customers Who Bought This Also Bought