VWR® Drosophila Vials, Narrow and Wide
Catalog #: 75813-144
Supplier: VWR International
Specifications
- Cap included:No
- Description:VWR® Drosophila Vial, Narrow
- Material:PP
- Packaging:Bulk
- Diameter ×H:25×95 mm
Specifications
About this item
Variety of sizes and materials for all applications.
- High quality, virgin grade plastic
- Wide and narrow opening diameters
- Tray packs enable easier food loading
VWR® Drosophila vials provide multiple options for storing fly cultures. Manufactured from pure virgin medical grade plastics, the vials are available in crystal clear polystyrene, translucent flexible and autoclavable polypropylene, or virtually unbreakable K-Resin. The flat bottom, straight wall design allows for convenient racking, stacking, and filling. The vials are available in bulk quantities, or in a 10 by 10 tray pack for quick and effortless filling. The bulk quantities are layered with open-end orientation for convenient handling and small case size.
Customers Who Bought This Also Bought