Order Entry
ContactUsLinkComponent
SLIDE MIC SUPERFROST+ CLPDCRNR WHT 3X1IN
Superfrost® Plus and Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Catalog #: 95057-985
Supplier:  Erie Scientific
SLIDE MIC SUPERFROST+ CLPDCRNR WHT 3X1IN
Superfrost® Plus and Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Catalog #: 95057-985
Supplier:  Erie Scientific

Specifications

  • Color:
    White
  • Description:
    Superfrost® Plus, Clipped Corner
  • L×W:
    75×25" mm (3×1")
  • Thickness:
    1 mm (1/32")
  • Cat. no.:
    95057-985
  • Supplier No.:
    4951PLUS-4

Specifications

About this item

Slides are treated to electrostatically attract frozen tissue sections and cytology preparations, eliminating the need for adhesives or coating steps.

Customers Who Bought This Also Bought