Components and Accessories for Miele Lab Washers, Miele Professional
Catalog #: CA75806-574
Supplier: Miele
Specifications
- Description:A150 Lower Injection Basket
- Cat. no.:CA75806-574
- For Use With:For Inject Modules Only
- Supplier No.:69615000D
- Details:—
Specifications
About this item
Various accessories.
Customers Who Bought This Also Bought