Components and Accessories for Miele Lab Washers, Miele Professional
Catalog #: CA75806-566
Supplier: Miele
Specifications
- Description:A102 Upper Basket with Open Front
- Cat. no.:CA75806-566
- For Use With:For Inserts for Test Tubes, Funnels, Beakers
- Supplier No.:69610200D
- Details:—
Specifications
About this item
Various accessories.
Customers Who Bought This Also Bought