Order Entry
ContactUsLinkComponent
PLATE ASY LOW FLNG TSU CULT TRTD 384WELL
Corning® Clear Bottom, Black and White, Polystyrene Microplates, 384 Well, Corning
Catalog #: 75805-274
Supplier:  Corning
   Low Manufacturing Impact
PLATE ASY LOW FLNG TSU CULT TRTD 384WELL
Corning® Clear Bottom, Black and White, Polystyrene Microplates, 384 Well, Corning
Catalog #: 75805-274
Supplier:  Corning
   Low Manufacturing Impact

Specifications

  • Sterility:
    Sterile
  • Well Volume:
    112 µl
  • Color:
    White with Clear Bottom
  • Description:
    Corning® 384-well, Polystyrene
  • Environmentally Preferable:
    Low Manufacturing Impact
  • No. of wells:
    384
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Cat. no.:
    75805-274
  • Packaging:
    20/Bag
  • Well Shape:
    Flat-Bottom
  • Surface Treatment:
    Tissue culture (TC)
  • Supplier No.:
    3765

Specifications

About this item

PS, flat bottom. Suitable for fluorescent and luminescent assays using either top or bottom detection plate readers.

Customers Who Bought This Also Bought