Order Entry
ContactUsLinkComponent
Sabdex (Sabouraud Dextrose) Agar with Chloramphenicol 15x100mm plate
Sabdex (Sabouraud Dextrose) Agar, Hardy Diagnostics
Catalog #: CA52428-858
CAS Number:  
Sabdex (Sabouraud Dextrose) Agar with Chloramphenicol 15x100mm plate
Sabdex (Sabouraud Dextrose) Agar, Hardy Diagnostics
Catalog #: CA52428-858
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Packaging:
    Plate
  • Description:
    With Chloramphenicol
  • Size:
    15x100mm
  • Storage temperature:
    2-8C
  • Cat. no.:
    CA52428-858
  • Fill Volume:
    26 ml
  • Application:
    For the selective isolation of fungi.
  • Supplier No.:
    W72

Specifications

About this item

Sabdex (Sabouraud Dextrose) Agar with Chloramphenicol, deep fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics

(Sabourauds Dextrose Agar with Chloramphenicol) For the selective isolation of fungi.

Customers Who Bought This Also Bought