VWR® Erlenmeyer Flasks, Wide Neck
Catalog #: 75804-646
Supplier: VWR International
Specifications
- Capacity:250 ml
- Cat. no.:75804-646
Specifications
About this item
Made of borosilicate glass 3.3.
Customers Who Bought This Also Bought
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
57,66 $ to 576,72 $
VWR® Volumetric Flask, Clear Glass, Wide Neck, Class A, Unserialized
Supplier: VWR International
396,37 $ to 720,56 $
VWR® Graduated Cylinders, Calibrated To Contain, Class B
Supplier: VWR International
84,94 $ to 508,22 $
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
205,73 $
CELLSTAR® Cell Culture Multiwell Plates, Polystyrene, Greiner Bio-One
Supplier: Greiner Bio-One
188,29 $
Acetic acid glacial 99.7-100.5% ACS, meets analytical specification of USP, FCC, VWR Chemicals BDH®
Supplier: BDH
496,17 $ to 2 375,72 $