Adapter 8 x 66 mm
Catalog #: 75801-846
Supplier: HETTICH INSTRUMENTS
Customers Who Bought This Also Bought
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Nitric acid 67 - 70%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Supplier: BDH
$729.52