Order Entry
ContactUsLinkComponent
Zobell solution OPR calibrator 250 ml
Zobell Solution ORP Calibrator, YSI
Catalog #: 75801-382
Supplier:  YSI
CAS Number:  
Zobell solution OPR calibrator 250 ml
Zobell Solution ORP Calibrator, YSI
Catalog #: 75801-382
Supplier:  YSI
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Zobell Solution ORP Calibrator
  • Size:
    250 mL
  • Cat. no.:
    75801-382
  • Supplier No.:
    061321

Specifications

About this item

Zobell solution is used for calibrating ORP/Redox on several different instruments.

  • Temperature chart for convenience
  • Simple calibration value inputs
  • Concentrated powder

A convenient temperature chart with corresponding ORP values is included for simple calibration value inputs. Concentrated powder makes 125, 250, or 500 mL of calibration standard. Zobell solution is used for calibrating ORP/Redox on several different instruments.

Packaging: Dated Material. Unmixed - Five Years. Mixed - Six Months
Customers Who Bought This Also Bought