ContactUsLinkComponent
Cap ValuLine F217 and PTFE Lined 48-400
White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalog #: 75799-980
Supplier:  Qorpak
Product available on GSA Advantage®
Cap ValuLine F217 and PTFE Lined 48-400
White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Catalog #: 75799-980
Supplier:  Qorpak
Product available on GSA Advantage®
Specifications
  • Cap Size:
    48-400
  • Cat. no.:
    75799-980
  • Supplier No.:
    239085
Specifications
About this item
Polypropylene caps with F217 & Fluoropolymer Resin liners are an economical alternative to Thermoset & Phenolic Fluoropolymer Resin lined caps.
  • Liners are very resilient – great sealing capability
  • Liners have strong resistance to chemicals and moisture
Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Customers Who Bought This Also Bought