Biox Cell Spreaders
Catalog #: 75799-958
Supplier: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Color:Orange
- Packaging:Individually Wrapped
- Supplier No.:65-1001-1
Specifications
About this item
Cell Spreaders are ideal for the even spreading of bacterial cultures.
- Polystyrene construction
- L-Shape design
- Disposable
- Smooth spreading surface with slight upward angle reduces the risk of damaging agar surfaces
- Gamma radiation sterilized
- Individually wrapped
L-Shaped cell spreaders allows for application on the entire culture dish or plate surface.
Customers Who Bought This Also Bought