Biox Inoculating Loops and Needles
Catalog #: 75799-954
Supplier: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Color:Orange
- Description:Loop, 10 µL
- Supplier No.:65-0010
Specifications
About this item
Innoculating loops and needles are smooth and flexible for uniform and smooth streaking, without damaging the gel surface.
- Sterile
- Polystyrene
- Other end of the loops can be used as a needle
- Free from lubricants, oils, and electrostatic charges
Needles are suitable for removal of single colony specimens and the opposite end of each loop can be used as a needle.
Customers Who Bought This Also Bought