Biox Petri Dishes
Catalog #: 75799-946
Supplier: TWD Scientific
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Ø:90 mm
- Height:15 mm
- Supplier No.:66-1501
Specifications
About this item
Petri Dishes are manufactured from crystal polystyrene for optimal analysis by microscopy.
- Easy stacking
- Triple vented lid for improved gas exchange
- Heavy design for optimal heat resistance
- Clear glass-like clarity
- Ethylene oxide sterilized
Highly transparent for microscopic analysis and vented lids for better gas exchange.
Customers Who Bought This Also Bought