AlphaTec® 87-086 Natural Rubber Gloves, Ansell
Catalog #: 75795-470
Supplier: Ansell Canada
Specifications
- Glove Color:Silver
- Glove size:8,5
- Length:305 mm (12")
- Latex/Latex-free:Latex
- Cuff style:Beaded
- Liner material:Flock
- Finish:Reversed lozenge
- Cat. no.:75795-470
- Supplier No.:113519
Specifications
About this item
Flocked, natural rubber gloves are comfortable and food compliant.
- Beaded cuff for tear resistance and easy donning
- Food approved
- Good abrasion resisstance
- 100% cotton flock lining
The AlphaTec® 87-086 natural rubber latex glove is flock lined and lightweight. It is resistant, flexible, and very comfortable. Ideally suited for use in low hazard areas.
Certifications: EN 420:2003 + A1:2009; FDA 21 CFR; CFIA.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.
Customers Who Bought This Also Bought