To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:3.8 L (1 gal.)
- Packaging:Bottle
- Sterility:Non sterile
- Concentration:0.52%
- Supplier No.:SHC-01-0.52
Specifications
About this item
HYPO-CHLOR® is an effective one step, ready-to-use product that is available in premixed concentrations of 5.25%, 0.52%, and 0.25%.
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Made with USP Water for Injection
- Filtered at 0.2 microns
- Sterility tested according to current USP compendium
- Assayed according to current USP compendium
- Completely traceable from start to finish
- Completely validated for sterility and shelf life
Sodium hypochlorite solutions, formulated with WFI, used to clean cleanrooms and controlled areas including those used in health care institutions, biopharmaceutical, medical device, and diagnostic manufacturing facilities. HYPO-CHLOR® Products were developed for use in cleaning rotation cycles for maintaining a clean environment
Sterile sodium hypochlorite solutions are filtered at 0.2µm, gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation
Product uses: Use on hard, non-porous, and inanimate surfaces, on materials such as plastic, glass, vinyl, glazed porcelain, and stainless steel. Can also be used on machinery, tools, tables, counters, laminar flow benches, floors, walls and carts.