To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Lysing Matrix A
- Size:50×2 ml
- Adapter compatibility:QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96)
- Instrument Compatibility:FastPrep-24; FastPrep 120; FastPrep-96; SuperFastPrep-2
- Tube compatibility:2 ml
- Key applications:Extractions of nucleic acids, proteins, metabolites and other small molecules from virtually any sample
- Application areas:Sample Preparation; Sample Lysis; Nucleic Acid Isolation & Purification; Molecular Biology; Gene Expression; Microbiological Analysis; Gene Function; Gene Regulation
- Material of construction:Matrix A, contains garnet matrix and one 1/4 inch ceramic sphere.
- Matrix type:A
- Molecule enriched:DNA; RNA; Proteins; Other small molecules
- Materials required (not supplied):FastPrep® Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
- Storage and handling:Store at Room Temperature (15...30 °C)
- Sample volume:up to 200 mg per prep
- Sample type:Skeletal Muscle; Lung; Heart; Bone; Seeds; Bacteria (Gram +/-); Yeast; Fungi; Spores; Plants; Fixed Samples; Sludge; Wastewater; Feces; Food; Roots; Soil; Animal Tissue
- Specific gravity:Zirconium sphere, 5.5 - 6.1; Garnet Flakes, 4.0 - 4.1
- Product type:Lab Essentials
- Cat. no.:75784-610
- Supplier No.:116910050
Specifications
About this item
Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA.
- Chemically inert
- Will not bind nucleic acids
- Used for all sample types except soil
Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA
Each impact-resistant 2 mL tube contains garnet matrix and one, 1/4 inch ceramic sphere. Extra 1/4 inch ceramic spheres are packaged separately. Lysing Matrix A tubes have orange caps and are found in the FastDNA™ and FastRNA™ SPIN Kits. Lysing Matrix A is used for all sample types except soil for the subsequent isolation of genomic DNA, RNA, functional proteins and other metabolites and small molecules. This grinding matrix is chemically inert and will not bind nucleic acids.
This grinding matrix is chemically inert and will not bind nucleic acids.