Order Entry
ContactUsLinkComponent
Water Sterile Solution 1Gal
Hydrogen Peroxide WFI Sterile Solutions, STERIS®
Catalog #: CA75780-046
Supplier:  Steris
Water Sterile Solution 1Gal
Hydrogen Peroxide WFI Sterile Solutions, STERIS®
Catalog #: CA75780-046
Supplier:  Steris
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    6% Hydrogen Peroxide WFI Sterile Solution
  • Size:
    3.78 L (1 gal.)
  • Sterility:
    Each Lot USP 71 Sterility Tested
  • Packaging:
    Double Bagged
  • Formulation:
    Contains Minimal Organic Carbon
  • Cat. no.:
    CA75780-046
  • Supplier No.:
    1S0508WR

Specifications

About this item

STERIS® Hydrogen Peroxide WFI Sterile Solutions are manufactured with United States Pharmcopeia quality Water For Injection (WFI) and Hydrogen Peroxide

Sterile filtered and filled in a ISO Class 5 aseptic environment, each package is double bagged for easy entry into controlled environments and aseptic core areas. These products have an extensive documentation package to meet validation objectives and are supported by the STERIS® Technical Services team for individualized application and validation assistance.

This low residue cleaner with minimal organic carbon is designed to remove organic and inorganic residues from product contact surfaces, room surfaces, equipment, gloves, etc.

Certifications: Solution is manufactured in an FDA cGMP, ISO 9001 and ISO 13485 Compliant Quality System environment.
Customers Who Bought This Also Bought