Order Entry
ContactUsLinkComponent
Hi Tech Hand Lotion
Staticide® ESD Hi-Tech Hand Lotion, ACL
Catalog #: 75780-064
Supplier:  ACL Staticide
Hi Tech Hand Lotion
Staticide® ESD Hi-Tech Hand Lotion, ACL
Catalog #: 75780-064
Supplier:  ACL Staticide

Specifications

  • Capacity:
    473 ml (16 oz.)
  • Description:
    Staticide® ESD Hi-Tech Hand Lotion with Pump
  • Cat. no.:
    75780-064
  • Supplier No.:
    7016

Specifications

About this item

Static-dissipative hand lotion is essential for replenishing vital moisture on the skin that is important for wrist strap performance

This static-dissipative hand lotion is equipped to prevent and handle static discharge, especially in the electronics industry.

Certifications: Complies with DOD 1686A and DOD HDBK 263.
Customers Who Bought This Also Bought