Neoprene Burner Tubing
Catalog #: 470220-806
Supplier: Wards
Specifications
- Description:Neoprene connecting hose, 90 cm
- Cat. no.:470220-806
- Supplier No.:CH0100B
Specifications
About this item
Connect your burner to a fuel source easily and safely.
- Two sizes available
- Tight secure connection to most common burners and gas sources
This neoprene hose connects your bunsen, tirrill or any other type of burner to a fuel source.
Customers Who Bought This Also Bought