Seam-Sealing Tape, ChemTape® PIG®, New Pig
Catalog #: 10860-942
Supplier: New Pig
Specifications
- Dimensions:2" W x 180' L
- Description:ChemTape®
- Cat. no.:10860-942
- Supplier No.:WPL591
Specifications
About this item
Seam-sealing tape designed and tested for use with chemical-resistant clothing.
- Ideal for hazmat response
- Designed and tested to ensure chemical resistance
- Excellent for securing gloves and boots
Ideal for hazmat response, this tape is designed and tested to ensure chemical resistance. Tears easily by hand and protects better than duct tape. Excellent for securing gloves and boots to protective clothing.
Customers Who Bought This Also Bought