Order Entry
ContactUsLinkComponent
Acid Encapsulating & Neutralizing Pillow
Acid Encapsulating & Neutralizing Pillow, PIG®
Catalog #: 10860-080
Supplier:  New Pig
Acid Encapsulating & Neutralizing Pillow
Acid Encapsulating & Neutralizing Pillow, PIG®
Catalog #: 10860-080
Supplier:  New Pig

Specifications

  • Description:
    PIG® Acid Encapsulating & Neutralizing Pillow
  • Cat. no.:
    10860-080
  • Absorbency:
    32 oz./pillow
  • Supplier No.:
    PIL352

Specifications

About this item

Encapsulate and neutralize the most common acid spills with the Acid Encapsulating & Neutralizing Pillow.

  • Great for battery charging
  • Identify best times for handling
  • Instantly absorbs up to 98% sulfuric acid conectration

Encapsulate and neutralize the most common acid spills; great for battery charging, transport and storage areas. Color changes as liquids are taken in, then returns to normal when neutralization is complete; helps identify the best time to handle with the least amount of risk. Instantly absorbs up to 98% concentrations of sulfuric acid.

Customers Who Bought This Also Bought