To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:475 ml (16 oz.)
- Cat. no.:CA89233-178
- Packaging:Poly Bottle
- Supplier No.:4101
Specifications
About this item
Conflikt ® Ready-to-use Disinfectant Spray cleans and disinfects hard surfaces that may harbor viruses, bacteria and fungi
Pre-diluted, stable solution of dual quaternary ammonium active ingredients is effective against HIV-1, E-coli and other pathogenic organisms. Conforms to OSHA's Bloodborne Pathogens Standard. Conflikt is effective against a wide range of organisms including TB. EPA registered.