Order Entry
ContactUsLinkComponent
VWR* Bootcover Medium
VWR® Disposable Sterile Boot Covers, SAL 10⁻⁶
Catalog #: 10858-530
Supplier:  VWR International
VWR* Bootcover Medium
VWR® Disposable Sterile Boot Covers, SAL 10⁻⁶
Catalog #: 10858-530
Supplier:  VWR International

Specifications

  • Garment application:
    Cleanroom,
    Electronics/semi-conductor,
    Laboratory,
    Pharmaceutical
  • Length:
    72.6 cm (28.6")
  • Shoe size:
    M
  • Color:
    White
  • Fabric:
    SMS/PVC
  • Cat. no.:
    10858-530

Specifications

About this item

Disposable sterile boot covers are manufactured from 60gsm SMS fabric, and feature a 250gsm nonskid PVC sole.

  • Disposable
  • Sterile
  • Vacuum packed in pairs

All boot covers feature bonded seams and elastic uppers for a secure fit. These garments are sterilized to a SAL of 10¯⁶. Boot covers are vacuum packed in pairs.

Certifications: ISO9001, ISO13485, ISO14001, CE, GMP.
Customers Who Bought This Also Bought