Grippy® Absorbent Mat Roll, PIG®
Catalog #: 10860-050
Supplier: New Pig
Specifications
- Description:PIG® Grippy® Absorbent Mat Roll
- Cat. no.:10860-050
- Absorbency:2.6 gal./roll
- Supplier No.:MAT1650
Specifications
About this item
The only absorbent mat with Grippy® backing stays put no matter what.
- Full-coverage adhesive backing holds tight but peels up easily
- Eliminate tapes and trays
- No curling or bunching
- Keeps floors completely dry
Specially formulated, full-coverage adhesive backing holds tight but peels up easily; eliminates the need for tapes and trays. No curling or bunching; lies flat to reduce trip hazards around workstations, walkways and machinery. Eight layers of fine-fiber polypropylene safely absorb leaks, drips and overspray of oils, coolants, solvents and water to keep liquids from being tracked around. Poly backing creates a liquid proof barrier that keeps floors completely dry.
Customers Who Bought This Also Bought