ContactUsLinkComponent
Merlin MicroShot Injector
Merlin MicroShot Injector, Restek
Catalog #: 10855-834
Supplier:  Restek
Merlin MicroShot Injector
Merlin MicroShot Injector, Restek
Catalog #: 10855-834
Supplier:  Restek
Specifications
  • Description:
    Merlin MicroShot Injector, 2.0 µL injection
  • Cat. no.:
    10855-834
  • Supplier No.:
    22230
Specifications
About this item
Increase the accuracy and reproducibility of manual injections with the Merlin MicroShot injector.
  • NIST traceability assures accurate injections
  • Fixed volume reduces sampling error
  • No need to transfer to autosampler vials
  • Prevents bent syringe plungers
  • Five injection volumes available
This new injector is calibrated to NIST reference standards to assure accurate and traceable displacement. Precise repeated injections of the preset volume can be made using a standard autosampler syringe with less variation than when injecting by hand. The trigger mechanism provides rapid sample delivery, which reduces needle residence time in the injection port and minimizes potential sample discrimination. The Merlin MicroShot injector allows convenient sampling from a wide variety of containers, so you can save time by eliminating the need to transfer aliquots into autosampler vials. The design of this unit also includes a plunger support, which protects the syringe plunger and prevents it from bending. Fits Agilent-style (ball-end plunger) autosampler syringes with either fixed or removable, 23- or 26-gauge needles.
Ordering information: Syringe not included. Requires Agilent-style (ball-end plunger) autosampler syringe.
Customers Who Bought This Also Bought