Inserts for Versa Vials™, Restek
Catalog #: 10854-362
Supplier: Restek
Specifications
- Cap included:No
- Capacity:0.25 ml
- Description:Versa vial insert with glass flange
- Material:Glass
- Cat. no.:10854-362
- Bottom Style:Conical bottom
- Supplier No.:22707
Specifications
About this item
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
- 250 µl volume vial compatibility
- Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
Customers Who Bought This Also Bought