Accessories for High-Power Electronic Crimping Tool, Restek
Catalog #: 10854-198
Supplier: Restek
Specifications
- Description:Interchangeable Crimper Jaw Set for High-Powered Electronic Crimping Tool
- Cat. no.:10854-198
- Seal Size:11 mm
- Supplier No.:23424