Magnetic Screw-Thread Caps, 18 mm, Restek
Catalog #: 10854-176
Supplier: Restek
Specifications
- Description:Magnetic Caps and Septa for SPME
- Cat. no.:10854-176
- Supplier No.:23091
- Orifice:18 mm
Specifications
About this item
18 mm magnetic screw-thread caps are quickly applied by magnetic autosamplers and feature wide orifices for ensuring efficient needle insertion without breaking.
- Economical
- Sturdy fabrication
- Wide orifice for easy needle insertion
These screw-thread caps are compatible with magnetic autosamplers, making application easier for the user.
Customers Who Bought This Also Bought