ContactUsLinkComponent
Valco* Internal Tee 0.75 mm
Zero-Dead-Volume Valco® Internal Tee, Restek
Catalog #: 10852-958
Supplier:  Restek
Valco* Internal Tee 0.75 mm
Zero-Dead-Volume Valco® Internal Tee, Restek
Catalog #: 10852-958
Supplier:  Restek
Specifications
  • Description:
    Internal Tee
  • Cat. no.:
    10852-958
  • Hole Size:
    0.75 mm
  • Supplier No.:
    20155
Specifications
About this item
Use these tee connectors for making three-way connections with tubing.
  • 300 series stainless steel; stainless steel ferrules included
  • For 1/16" OD tubing
Designed so that no volume is introduced by the fitting which is not cleanly swept.
Customers Who Bought This Also Bought