High Five Industrial Grade Disposable Vinyl Gloves, Powder-Free, Microflex®, Ansell
Catalog #: CA89427-524
Supplier: Microflex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Glove Color:Translucent
- Glove size:S
- Disposable / reusable:Disposable
- Thickness:4 mil
- Cat. no.:CA89427-524
- Supplier No.:V281
Specifications
About this item
Industrial-grade disposable vinyl gloves are suitable for general-purpose applications that do not require medical-grade protection.
- Outstanding elasticity and tensile strength
- Powder-free
Great for applications that do not require a medical-grade glove.
Customers Who Bought This Also Bought