Corning® Gosselin™ Petri Dishes, Corning
Catalog #: 10852-282
Supplier: Corning
Specifications
- Ø:90 mm
- Sterility:Sterile
- Description:Round Petri Dish with 3 Vents
- Height:14.2 mm
- Packaging:33/Bag, 825 Bags/Case
- Supplier No.:BP93B-102
Specifications
About this item
Contact plates used in hygiene control within the pharmaceutical or food processing industry, as well as in hospital environments. Manufactured in a Class 100 environment with cleanroom packaging. Available aseptic or sterile (SAL 10⁻⁶).
- Moulded grid on the bottom makes it possible to identify contamination per cm²
- Grip lid ensures safety during transport
- Domed base guarantees a saving in agar whilst maintaining safe sampling
- Stable stacking
The Petri Dishes are packaged with easily accessible traceability features such as peelable labels with lot numbers, valid expiration date and sterilization information on each case. It is used in industrial microbiology labs such as Food and Beverage, Environmental/Water Testing, etc.
They are made of crystal polystyrene and are optimized for automation.
Customers Who Bought This Also Bought