ContactUsLinkComponent
Sheep Brain, Preserved, Injection: Plain, without hypophysis
Ward's® Fully Extracted Sheep Brains (Dura Mater Removed)
Catalog #: 470220-070
Supplier:  Cyrgus
Sheep Brain, Preserved, Injection: Plain, without hypophysis
Ward's® Fully Extracted Sheep Brains (Dura Mater Removed)
Catalog #: 470220-070
Supplier:  Cyrgus
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Cat. no.:
    470220-070
  • Packaging:
    Vacuum pack of 10
  • Preservative:
    Formalin preserved
Specifications
About this item
Formalin preserved sheep brains with dura mater removed. Available in jars or pails. Shipped in nontoxic Wardsafe®.
  • Dura mater removed
  • Brain anatomy
These fully extracted sheep brains have the hypophysis and cranial nerves intact. Sheep brains are great for physiological study and exploring neurological anatomy that is similar to the human brain.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."
Customers Who Bought This Also Bought