Type I Poly Safety Cans, Justrite®
Catalog #: 10836-908
Supplier: Justrite
Specifications
- Capacity:1 gal.
- Description:Safety Can, 1G White Oval Poly
- Cat. no.:10836-908
- Supplier No.:12162
Specifications
About this item
Chemical-resistance HDPE cans offer safe storage for flammable corrosives.
Certifications: NFPA, OSHA.
Customers Who Bought This Also Bought