Order Entry
ContactUsLinkComponent
VWR Collection Striped Safety Tape 2in
VWR® Striped Safety Tape
Catalog #: 10841-232
Supplier:  VWR International
VWR Collection Striped Safety Tape 2in
VWR® Striped Safety Tape
Catalog #: 10841-232
Supplier:  VWR International

Specifications

  • Length:
    18 yds.
  • Width:
    2"
  • Description:
    Black/Yellow Stripe
  • Cat. no.:
    10841-232
  • Supplier No.:
    VWSST-2BLYE

Specifications

About this item

Striped pressure sensitive tape for delineating and color coding specific equipment and areas.

  • Promotes plant safety
  • Can be used for marking emergency exits or safety equipment

This laminated vinyl tape is wound on a plastic core, is scuff resistant and holds up well to solvents, humidity and oil. Available in black/yellow, blue/white and red/white, SST can be used for marking fire equipment/exits, hazardous areas and first aid points within cleanrooms and controlled environments.

Customers Who Bought This Also Bought