Versaflo™ S-Series Economy Hoods, 3M™
Catalog #: CA89205-950
Supplier: 3M
Specifications
- Color:White
- Description:Economy Hood
- Size:Medium-Large
- Supplier No.:S-403L-20
Specifications
About this item
Helps provide head, face, neck and shoulder coverage.
- Provides head, face, neck and shoulder coverage
- For applications that require frequent replacement of entire headgear assembly
- Fully assembled
- OSHA APF 1000
Can be suitable for applications that require frequent replacement of the entire headgear assembly, such as pharmaceutical manufacturing, or in situations where there is less soiling of the outer fabric.
Fully assembled and ready to use out of the box. Color: white.
Certifications: OSHA APF 1000
Ordering information: Includes hood, suspension, and inner shroud.
Customers Who Bought This Also Bought